Learn More
Sino Biological Protein Human CA4 Biotin 100UG
Catalog No. 5001073634
Encompass_Preferred
A DNA sequence encoding the Human Carbonic Anhydrase IV/CA4 (NP 000708 1) (Ala19-Lys283) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-73634 Supplier Sino Biological Supplier No. 10472H49HB100
A DNA sequence encoding the Human Carbonic Anhydrase IV/CA4 (NP 000708 1) (Ala19-Lys283) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed