Learn More
Sino Biological Recombinant Human CD55/DAF Protein (His Tag), Biotinylated, HPLC-verified 20 µg
Catalog No. 5000656565
Encompass_Preferred
A DNA sequence encoding the human CD55 precursor (NP_000565.1) (Met 1-Ser 353) was expressed, with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-006-56565 Supplier Sino Biological Supplier No. 10101H08HB20
A DNA sequence encoding the human CD55 precursor (NP_000565.1) (Met 1-Ser 353) was expressed, with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.