Learn More
Sino Biological Recombinant Human Cadherin-16 Protein (His Tag), Biotinylated 100 µg
Catalog No. 5000656529
Encompass_Preferred
A DNA sequence encoding the Human Cadherin-16 (NP_004053.1) (Met1-Ala786) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-006-56529 Supplier Sino Biological Supplier No. 10915H08HB100
A DNA sequence encoding the Human Cadherin-16 (NP_004053.1) (Met1-Ala786) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.