Learn More
Sino Biological Protein Human CDK4 CCND3 10UG
Catalog No. 5001073900
Encompass_Preferred
Recombinant full-length human CDK4 and CyclinD3 (S259A) were co-expressed by baculovirus in Sf9 cells using an N-terminal GST tag on both proteins
Catalog No. 50-010-73900 Supplier Sino Biological Supplier No. C31128G10
Recombinant full-length human CDK4 and CyclinD3 (S259A) were co-expressed by baculovirus in Sf9 cells using an N-terminal GST tag on both proteins