Learn More
Sino Biological Protein Human CDK6 CCND3 10UG
Catalog No. 5001074150
Encompass_Preferred
Recombinant full-length human CDK6 and CyclinD3 (S259A) were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins
Catalog No. 50-010-74150 Supplier Sino Biological Supplier No. C35128G10
Recombinant full-length human CDK6 and CyclinD3 (S259A) were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins