Learn More
BioVendor, LLC Recombinant proteinTotal 438 AA, M.w. 51.27kDa (calculated)
SDP
Supplier:  BioVendor, LLC RD172034100
Encompass
Recombinant proteinTotal 438 AA, M.w. 51.27kDa (calculated). The AA sequence (AA 1¿427) is identical to Swiss-Prot-P10909 (AA 23¿449, secreted Human Clusterin). C-terminal Flag-tag, 11 extra AA (highlighted).Amino Acid SequenceDQTVSDNELQEMSNQGSKYVNKEIQNAVNGVKQIKTLIEKTNEERKTLLSNLEEAKKKKEDALNETRESETKLKELPGVCNETMMALWEECKPCLKQTCMKFYARVCRSGSGLVGRQLEEFLNQSSPFYFWMNGDRIDSLLENDRQQTHMLDVMQDHFSRASSIIDELFQDRFFTREPQDTYHYLPFSLPHRRPHFFFPKSRIVRSLMPFSPYEPLNFHAMFQPFLEMIHEAQQAMDIHFHSPAFQHPPTEFIREGDDDRTVCREIRHNSTGCLRMKDQCDKCREILSVDCSTNNPSQAKLRRELDESLQVAERLTRKYNELLKSYQWKMLNTSSLLEQLNEQFNWVSRLANLTQGEDQYYLRVTTVASHTSDSDVPSGVTEVVVKLFDSDPITVTVPVEVSRKNPKFMETVAEKALQEYRKKHREEAAADYKDDDDKAsp1toThr3andSer5wereconfirmedbyN-terminalsequencing.SourceHEK293PurityPurity as determined by densitometric image analysis: >90% Endotoxin< 1.0 EU/ugApplicationsWestern blotting, ELISA, Cell culture and/or animal studies
Catalog No. 50-871-248