Learn More
Sino Biological Recombinant Human CTLA-4 Protein (His & AVI Tag), Avi-tag Biotinylated, HPLC-verified 20 µg
Catalog No. 5000656754
Encompass_Preferred
A DNA sequence encoding the Human CTLA-4 (NP_005205.2) (Lys36-Phe162) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-006-56754 Supplier Sino Biological Supplier No. 11159H49H1B20
A DNA sequence encoding the Human CTLA-4 (NP_005205.2) (Lys36-Phe162) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.