Learn More
BioVendor, LLC PROTEIN HUMAN FGF-19 0.1 MG
SDP
Supplier:  BioVendor, LLC RD172107100
Encompass
Recombinant protein Description Total 206 AA. Mw: 23 kDa (calculated). UniProtKB acc.no. O95750. N-terminal His-tag, 14 extra AA (highlighted).Amino Acid SequenceMRGSHHHHHHGMASLAFSDAGPHVHYGWGDPIRLRHLYTSGPHGLSSCFLRIRADGVVDCARGQSAHSLLEIKAVALRTVAIKGVHSVRYLCMGADGKMQGLLQYSEEDCAFEEEIRPDGYNVYRSEKHRLPVSLSSAKQRQLYKNRGFLPLSHFLPMLPMVPEEPEDLRGHLESDMFSSPLETDSMDPFGLVTGLEAVRSPSFEKSource E. coliPurity >95% Endotoxin < 1.0 EU/ugFormulationFiltered (0.4 μm) and lyophilized in 0.5 mg/mL in 20mM TRIS, 50mM NaCl, pH 7.5ApplicationsWestern blotting, ELISAQuality Control TestBCA to determine quantity of the protein.SDS PAGE to determine purity of the protein.LAL to determine quantity of endotoxin.NoteThis product is intended for research use only.
Catalog No. 50-871-255