Learn More
Sino Biological Protein Human FGFR3 500UG
Catalog No. 5001074058
Encompass_Preferred
Recombinant human FGFR3 (397-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal TST-Trx-TEV tag
Catalog No. 50-010-74058 Supplier Sino Biological Supplier No. F0611T500
Recombinant human FGFR3 (397-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal TST-Trx-TEV tag