Learn More
Sino Biological Recombinant Human Glypican 4/GPC4 Protein (His & AVI Tag), Avi-tag Biotinylated, HPLC-verified 20 µg
Catalog No. 5000656806
Encompass_Preferred
A DNA sequence encoding the Human Glypican 4/GPC4 (O75487-1) (Ala19-Ser529) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-006-56806 Supplier Sino Biological Supplier No. 10090H49CB20
A DNA sequence encoding the Human Glypican 4/GPC4 (O75487-1) (Ala19-Ser529) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.