Learn More
Sino Biological Protein Human HMGB1 100UG
Catalog No. 5000699024
Encompass_Preferred
A DNA sequence encoding the Human HMGB1 (NP_002119.1) (Gly2-Glu215) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-99024 Supplier Sino Biological Supplier No. 10326H08H1UE100
A DNA sequence encoding the Human HMGB1 (NP_002119.1) (Gly2-Glu215) was expressed with a polyhistidine tag at the C-terminus.