Learn More
Sino Biological Protein Human HMGB1 1MG
Catalog No. 5000699085
Encompass_Preferred
A DNA sequence encoding the Human HMGB1 (NP_002119.1) (Gly2-Glu215) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-99085 Supplier Sino Biological Supplier No. 10326H08H1UE1
A DNA sequence encoding the Human HMGB1 (NP_002119.1) (Gly2-Glu215) was expressed with a polyhistidine tag at the C-terminus.