Learn More
Sino Biological Protein Human IL18 Biotin 20UG
Catalog No. 5001073818
Encompass_Preferred
A DNA sequence encoding the Human IL-18 (Q14116) (Tyr37-Asp193) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-73818 Supplier Sino Biological Supplier No. 10119H49EB20
A DNA sequence encoding the Human IL-18 (Q14116) (Tyr37-Asp193) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed