Learn More
Sino Biological Recombinant Human ST2/IL-1 RL1 Protein (His Tag), Biotinylated, HPLC-verified 20 µg
Catalog No. 5000656745
Encompass_Preferred
A DNA sequence encoding the human IL1RL1 isoform 2 (NP_003847.2) (Met1-Phe328) was expressed fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-006-56745 Supplier Sino Biological Supplier No. 10105H08HB20
A DNA sequence encoding the human IL1RL1 isoform 2 (NP_003847.2) (Met1-Phe328) was expressed fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.