Learn More
Sino Biological Recombinant Human IL-1RA/IL-1RN Protein, Biotinylated 100 µg
Catalog No. 5000656747
Encompass_Preferred
A DNA sequence encoding the mature form of human IL1RA isoform1 (NP_776214.1) (Arg 26-Glu 177) was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-006-56747 Supplier Sino Biological Supplier No. 10123HNAEB100
A DNA sequence encoding the mature form of human IL1RA isoform1 (NP_776214.1) (Arg 26-Glu 177) was expressed. The purified protein was biotinylated in vitro.