Learn More
Sino Biological Recombinant Human IL-25/IL-17E Protein (hFc Tag), Biotinylated 100 µg
Catalog No. 5000656546
Encompass_Preferred
A DNA sequence encoding the mature form of human IL25 (Tyr 33-Gly177) (NP_073626.1) was fused with the Fc region of human IgG1 at the N-terminus via a polypeptide linker. The purified protein was biotinylated in vitro.
Catalog No. 50-006-56546 Supplier Sino Biological Supplier No. 10096H01HB100
A DNA sequence encoding the mature form of human IL25 (Tyr 33-Gly177) (NP_073626.1) was fused with the Fc region of human IgG1 at the N-terminus via a polypeptide linker. The purified protein was biotinylated in vitro.