Learn More
Sino Biological Protein Human IL6R IL6 1MG
Catalog No. 5000698640
Encompass_Preferred
A DNA sequence encoding the Human IL-6/IL-6R alpha Chimera (P08887 & P05231) (Leu20-Asp358 & Val30-Met212) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-006-98640 Supplier Sino Biological Supplier No. CC001H07H1UE1
A DNA sequence encoding the Human IL-6/IL-6R alpha Chimera (P08887 & P05231) (Leu20-Asp358 & Val30-Met212) was expressed, with a polyhistidine tag at the N-terminus.