Learn More
Sino Biological Protein Human KDR AF488 25UG
Supplier:  Sino Biological 10012H86HSD25
Encompass_Preferred
A DNA sequence encoding the human VEGFR2 (NP 002244 1) (Met 1-Glu 764) was expressed with a C-terminal polyhistidine tag The protein is site-specifically conjugated with AF 488 (Excitation Max 495 nm Emission Max 519 nm)
Catalog No. 50-010-73819