Learn More
Sino Biological Protein Human KDR AF647 100UG
Catalog No. 5001074038
Encompass_Preferred
A DNA sequence encoding the human VEGFR2 (NP 002244 1) (Met 1-Glu 764) was expressed with a C-terminal polyhistidine tag The protein is site-specifically conjugated with AF 647 (Excitation Max 655 nm Emission Max 680 nm)
Catalog No. 50-010-74038 Supplier Sino Biological Supplier No. 10012H86HSG100
A DNA sequence encoding the human VEGFR2 (NP 002244 1) (Met 1-Glu 764) was expressed with a C-terminal polyhistidine tag The protein is site-specifically conjugated with AF 647 (Excitation Max 655 nm Emission Max 680 nm)