Learn More
Sino Biological Protein Human MFGE8 100UG
Catalog No. 5001073855
Encompass_Preferred
A DNA sequence encoding the Human MFGE8 (Q08431) fragment (Leu24-Thr68 linked to Glu226-Cys387) was fused with the Fc region of human IgG1 (H229Y) at the C-terminus
Catalog No. 50-010-73855 Supplier Sino Biological Supplier No. 10853H02H100
A DNA sequence encoding the Human MFGE8 (Q08431) fragment (Leu24-Thr68 linked to Glu226-Cys387) was fused with the Fc region of human IgG1 (H229Y) at the C-terminus