Learn More
Sino Biological Recombinant Human NCAM-1/CD56 Protein (Site-Specific AF 488 Conjugation, His Tag) 100 µg
Catalog No. 5000656748
Encompass_Preferred
A DNA sequence encoding the human NCAM1 (NP_001070150.1) (Met 1-Pro 603) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with AF 488 (Excitation Max.= 495 nm, Emission Max.= 519 nm).
Catalog No. 50-006-56748 Supplier Sino Biological Supplier No. 10673H86HSD100
A DNA sequence encoding the human NCAM1 (NP_001070150.1) (Met 1-Pro 603) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with AF 488 (Excitation Max.= 495 nm, Emission Max.= 519 nm).