Learn More
Sino Biological Recombinant Human NCAM-1/CD56 Protein (Site-Specific PE Conjugation, His Tag) 25 Tests
Catalog No. 5000656507
Encompass_Preferred
A DNA sequence encoding the human NCAM1 (NP_001070150.1) (Met 1-Pro 603) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with PE (Excitation= 495 - 566 nm, Emission Max.= 575 nm).
Catalog No. 50-006-56507 Supplier Sino Biological Supplier No. 10673H86HSP25
A DNA sequence encoding the human NCAM1 (NP_001070150.1) (Met 1-Pro 603) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with PE (Excitation= 495 - 566 nm, Emission Max.= 575 nm).