Learn More
Sino Biological Protein Human NRG1 ET-FREE 1MG
Catalog No. 5001074039
Encompass_Preferred
A DNA sequence encoding the N-terminal fragment (Ser 2-Lys 246) of human NRG1 isoform beta1 (Q02297-6) was fused with the Fc region of human IgG1 at the N-terminus
Catalog No. 50-010-74039 Supplier Sino Biological Supplier No. 11609H01H2UE1
A DNA sequence encoding the N-terminal fragment (Ser 2-Lys 246) of human NRG1 isoform beta1 (Q02297-6) was fused with the Fc region of human IgG1 at the N-terminus