Learn More
BioVendor, LLC Recombinant protein Total 671 AA. MW: 75 kDa (calculated)
SDP
Supplier:  BioVendor, LLC RD172045100
Encompass
Recombinant protein Total 671 AA. MW: 75 kDa (calculated). UniProtKB acc.no. Q15063. N-Terminal HisTag and Xa ¿ cleavage site 23 AA (highlighted).Amino Acid SequenceMGHHHHHHHHHHSSGHIEGRHMRNNHYDKILAHSRIRGRDQGPNVCALQQILGTKKKYFSTCKNWYKKSICGQKTTVLYECCPGYMRMEGMKGCPAVLPIDHVYGTLGIVGATTTQRYSDASKLREEIEGKGSFTYFAPSNEAWDNLDSDIRRGLESNVNVELLNALHSHMINKRMLTKDLKNGMIIPSMYNNLGLFINHYPNGVVTVNCARIIHGNQIATNGVVHVIDRVLTQIGTSIQDFIEAEDDLSSFRAAAITSDILEALGRDGHFTLFAPTNEAFEKLPRGVLERFMGDKVASEALMKYHILNTLQCSESIMGGAVFETLEGNTIEIGCDGDSITVNGIKMVNKKDIVTNNGVIHLIDQVLIPDSAKQVIELAGKQQTTFTDLVAQLGLASALRPDGEYTLLAPVNNAFSDDTLSMVQRLLKLILQNHILKVKVGLNELYNGQILETIGGKQLRVFVYRTAVCIENSCMEKGSKQGRNGAIHIFREIIKPAEKSLHEKLKQDKRFSTFLSLLEAADLKELLTQPGDWTLFVPTNDAFKGMTSEEKEILIRDKNALQNIILYHLTPGVFIGKGFEPGVTNILKTTQGSKIFLKEVNDTLLVNELKSKESDIMTTNGVIHVVDKLLYPADTPVGNDQLLEILNKLIKYIQIKFVRGSTFKEIPVTVYSourceE. coliPurity>90% Endotoxin< 0.1 EU/?g
Catalog No. 50-871-269