Learn More
Sino Biological Recombinant Human EPCR Protein (His Tag), Biotinylated, HPLC-verified 100 µg
Catalog No. 5000656531
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human PROCR (Q9UNN8) (Met 1-Thr 209), was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-006-56531 Supplier Sino Biological Supplier No. 13320H08HB100
A DNA sequence encoding the extracellular domain of human PROCR (Q9UNN8) (Met 1-Thr 209), was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.