Learn More
Sino Biological Recombinant Human RIOK2 Protein (Full length, GST Tag), Active 500 µg
Catalog No. 5000656593
Encompass_Preferred
Recombinant full-length human RIOK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-006-56593 Supplier Sino Biological Supplier No. R0510G500
Recombinant full-length human RIOK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.