Learn More
Sino Biological Protein Human TF ET-FREE 1MG
Catalog No. 5001073636
Encompass_Preferred
A DNA sequence encoding the Human Transferrin / TF (NP 001054 1) (Met1-Pro698) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-73636 Supplier Sino Biological Supplier No. 11019H08HUE1
A DNA sequence encoding the Human Transferrin / TF (NP 001054 1) (Met1-Pro698) was expressed with a polyhistidine tag at the C-terminus