Learn More
Sino Biological Protein Human TGFB1 500UG
Catalog No. 5000699388
Encompass_Preferred
A DNA sequence encoding the full length of human TGFβ1 (NP_000651.3) (Met 1-Ser 390) was fused with a C-terminal polyhistidine tag.
Catalog No. 50-006-99388 Supplier Sino Biological Supplier No. 10804H08HUE500
A DNA sequence encoding the full length of human TGFβ1 (NP_000651.3) (Met 1-Ser 390) was fused with a C-terminal polyhistidine tag.