Learn More
Sino Biological Recombinant Human TLR8 Protein (hFc Tag) 100 µg
Catalog No. 5000656516
Encompass_Preferred
A DNA sequence encoding the Human TLR8 (Q9NR97) (Glu27-Thr827) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-006-56516 Supplier Sino Biological Supplier No. 11914H02B100
A DNA sequence encoding the Human TLR8 (Q9NR97) (Glu27-Thr827) was expressed, fused with the Fc region of human IgG1 at the C-terminus.