Learn More
Sino Biological Protein Human TNFRSF13C 1MG
Catalog No. 5001073742
Encompass_Preferred
A DNA sequence encoding the Human BAFFR/TNFRSF13C (Q5H8V1) (Ser7-Ala71) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-010-73742 Supplier Sino Biological Supplier No. 16079H08H31
A DNA sequence encoding the Human BAFFR/TNFRSF13C (Q5H8V1) (Ser7-Ala71) was expressed with a C-terminal polyhistidine tag