Learn More
Sino Biological Protein Human VTN ET-FREE 1MG
Catalog No. 5001074049
Encompass_Preferred
A DNA sequence encoding the human VTN (NP 000629 3) (Met1-Leu478) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-74049 Supplier Sino Biological Supplier No. 10424H08HUE1
A DNA sequence encoding the human VTN (NP 000629 3) (Met1-Leu478) was expressed with a polyhistidine tag at the C-terminus