Learn More
Sino Biological Protein IL13 ET-FREE 500UG
Catalog No. 5001073864
Encompass_Preferred
A DNA sequence encoding the Human IL13 (AAK53823 1) (Met1-Asn132) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-73864 Supplier Sino Biological Supplier No. 10369H08HUE500
A DNA sequence encoding the Human IL13 (AAK53823 1) (Met1-Asn132) was expressed with a polyhistidine tag at the C-terminus