Learn More
Sino Biological Protein IL1B Biotin 100UG
Catalog No. 5001073938
Encompass_Preferred
A DNA sequence encoding the Human IL-1 beta (NP 000567 1) (Ala117-Ser269) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-73938 Supplier Sino Biological Supplier No. 10139H49EB100
A DNA sequence encoding the Human IL-1 beta (NP 000567 1) (Ala117-Ser269) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed