Learn More
Sino Biological Protein IL1RL2 ET-FREE 1MG
Catalog No. 5001074128
Encompass_Preferred
A DNA sequence encoding the human IL1RL2 (NP 003845 2) (Met1-Tyr337) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-74128 Supplier Sino Biological Supplier No. 14398H08HUE1
A DNA sequence encoding the human IL1RL2 (NP 003845 2) (Met1-Tyr337) was expressed with a polyhistidine tag at the C-terminus