Learn More
Sino Biological Protein IL6ST ET-FREE 100UG
Catalog No. 5001074051
Encompass_Preferred
A DNA sequence encoding the human IL6ST (AAA59155 1) (Met1-Ile618) was expressed with the Fc region of human IgG1 at the C-terminus
Catalog No. 50-010-74051 Supplier Sino Biological Supplier No. 10974H02HUE100
A DNA sequence encoding the human IL6ST (AAA59155 1) (Met1-Ile618) was expressed with the Fc region of human IgG1 at the C-terminus