Learn More
GenDEPOT Protein Isolation 10x1ml
Catalog No. 501959718
Encompass_Preferred
The Xpert Phosphatase Inhibitor Cocktail Solution(100X) is a mixture of several inhibitors to prevent valuable proteins from dephospholylation.
Catalog No. 50-195-9718 Supplier GenDEPOT Supplier No. P3200010
The Xpert Phosphatase Inhibitor Cocktail Solution(100X) is a mixture of several inhibitors to prevent valuable proteins from dephospholylation.