Learn More
Enzo Life Sciences Protein kinase C (19-31) (1mg). CAS: 121545-65-1
SDP
Supplier:  Enzo Life Sciences BMLP2060001
Encompass_Preferred
Shorter and more potent inhibitor of PKC (IC50=100 nM) than Protein Kinase C (19-36), Pseudosubstrate Inhibitor (BML-P150). Alternative name: PKC (19-31). Purity: ≥97% (HPLC). Formulation: TFA salt. Lyophilized. Solubility: Soluble in 5% acetic acid or in water. Long Term Storage: -20°C.
Catalog No. 50-201-1283