Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Protein Kinase C-epsilon Peptide Substrate

Catalog No. AAJ66938MCR
Change view
Click to view available options
Substrate Quantity:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Substrate Quantity
AAJ66938MCR 1mg
AAJ66938EXD 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66938MCR Supplier Thermo Scientific™ Supplier No. J66938MCR

missing translation for 'validMainframeMsg'

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Form Lyophilized
Molecular Weight (g/mol) 2067.45
Storage Requirements -30° to -10°C
Substrate Quantity 1mg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.