Learn More
CPC Scientific H-Ala-Gly-Asn-Lys-Val-Ile-Ser-Pro-Ser-Glu-Asp-Arg-Arg-Gln-Cys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PKCP034A
Encompass
ONE-LETTER SEQUENCE: AGNKVISPSEDRRQCMOLECULAR FORMULA: C66H114N24O24S1MOLECULAR WEIGHT:1659.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [159939-84-1], RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: M. Makowske et al., J. Biol. Chem., 263, 3402 (1988); B.K. McLLroy et al., Anal. Biochem., 195, 148 (1991)
Catalog No. 50-211-0184