Learn More
CPC Scientific H-Arg-Phe-Ala-Arg-Lys-Gly-Ala-Leu-Arg-Gln-Lys-Asn-Val-His-Glu-Val-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PKCP030A
Encompass
ONE-LETTER SEQUENCE: RFARKGALRQKNVHEVKMOLECULAR FORMULA: C89H153N33O22MOLECULAR WEIGHT:2037.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0182