Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protein kinase G inhibitor-1 | 354544-70-0 | 98.0% | C14H18N2O2S | 1 MG
SDP

Catalog No. 5000357771
Encompass_Preferred

Protein kinase G inhibitor-1 (Compound 270) is a mycobacterial protein kinase G inhibitor, with an IC50 of 0.9 μM. It can be used for mycobacterial infection research.

  • Chemical formula: C14H18N2O2S
  • CAS number: 354544-70-0
  • Purity: 98.0%

Catalog No. 50-003-57771 Supplier Medchemexpress LLC Supplier No. HY489581MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protein kinase G inhibitor-1 (Compound 270) is a mycobacterial protein kinase G inhibitor, with an IC50 of 0.9 μM. It can be used for mycobacterial infection research.

  • Chemical formula: C14H18N2O2S
  • CAS number: 354544-70-0
  • Purity: 98.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.