Learn More
Medchemexpress LLC HY-U00439A 5mg Medchemexpress, Protein kinase inhibitors 1 hydrochloride CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU00439A5MG
Encompass_Preferred
Medchemexpress, HY-U00439A 5mg Protein kinase inhibitors 1 hydrochloride CAS: Protein kinase inhibitors 1 hydrochloride is a potent HIPK2 inhibitor, with IC 50 s of 136 and 74 nM for HIPK1 and HIPK2, and a K d of 9.5 nM for HIPK2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4450