Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protein kinase inhibitor 6 | 348-45-8 | 99.04% | C13H9FN2S | 5 MG
SDP

Catalog No. 5000357880
Encompass_Preferred

Protein kinase inhibitor 6 is a protein kinase inhibitor. This product is for research purposes only and not for patient use.

  • Protein kinase inhibitor
  • Appearance: solid, white to off-white
  • Solubility in DMSO: 125 mg/mL
  • For research purposes only

Catalog No. 50-003-57880 Supplier Medchemexpress LLC Supplier No. HY470265MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Protein kinase inhibitor 6 is a protein kinase inhibitor. This product is for research purposes only and not for patient use.

  • Protein kinase inhibitor
  • Appearance: solid, white to off-white
  • Solubility in DMSO: 125 mg/mL
  • For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.