Learn More
ACROBiosystems Protein L-coupled Magnetic Beads (recommended for MPCLIA)
Supplier:  ACROBiosystems MPCA008100MG
Encompass_Preferred
The Protein L-coupled Magnetic Beads are 2.8 µm superparamagnetic particles covalently coupled to a highly pure form of Protein L. The beads can be used to capture the antibodies in Chemiluminescence procedures. The Protein L is a 45 kDa surface protein, and can bind almost all antibody types, including IgG, IgM, IgA, IgE and IgD. It binds to antibodies with appropriate Kappa light chains such as VkI, VkIII and VkIV, whereas no binding occurred with antibodies of the VkII or with any Lambda light chains. The Protein L-coupled Magnetic Beads is easy to capture the most antibodies, and the bounded antibody have no activity lost, this ready to use products could greatly save your protein coupling time and hassle, and help us get the best performance and highly reproducible results.
Catalog No. 50-233-1953