Learn More
CELLTECHGEN Protein L, cl-APC conjugate, 100 ug
Supplier:  CELLTECHGEN AXAP01012A
Encompass_Preferred
Despite this wide-ranging binding capability with respect to Ig classes, Protein L is not a universal immunoglobilin-binding protein. Binding of Protein L to immunoglobulins is restricted to those containing kappa light chains (i.e., k chain of the VL domain). Besides antibody, protein L is also suitable for binding of a wide range of antibody fragments such as Fabs, single-chain variable fragments (scFv), and domain antibodies.
Catalog No. NC1410279