The resDetect Protein L ELISA Kit is a sandwich ELISA for quantitative determination of recombinant Protein L in bioprocess in-process streams and final products. Supplied as a 96-test (8x12 strips) plate kit, it supports purification process development and routine quality control and is for research use only (RUO).
- Sandwich ELISA for quantitative measurement of Protein L.
- 96-test format (8x12 strips) suitable for routine assays.
- Sensitivity below 50 pg/mL with standard curve range 50 pg/mL-3,200 pg/mL.
- Sample volume 50 μL per well.
- Assay time approximately 2 hours.
- Includes pre-coated microplate, recombinant standard, biotin-conjugated detection antibody, streptavidin-HRP, buffers, substrate, and stop solution.
- Store unopened kit at 2-8 °C; opened components usable for 30 days per component instructions.
- For research use only (RUO); not for diagnostic use.