Learn More
Aniara Diagnostica LLC PROTEIN L LIGAND LEAKAGE ELISA
Supplier:  Aniara Diagnostica LLC A100027
Encompass
Medicago Protein L Ligand Leakage ELISA kit measures ligand leakage in eluates from Protein L affinity chromatography medium containing immunoglobulin (Ig) or Ig-fragments. Ready-to-use. Store components at 4C; Store reference at <-20C.
Catalog No. 50-100-8818