Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Protein L, PE Conjugate, Flow Cytometry, 100 μL
SDP

Catalog No. 502732812
Encompass

Protein L is a recombinant protein that interacts with the κ light chain variable domain of immunoglobulins without disrupting paratope function. Conjugated to phycoerythrin (PE), this product is specifically validated for direct flow cytometric analysis of human cells expressing κ light chain-containing immunoglobulins, including Fab fragments and single chain variable fragments (scFvs). The protein has been genetically engineered to remove cell wall, cell membrane, and albumin-binding regions to enhance specificity.

  • Validated for direct flow cytometry (live)
  • Recommended dilution: 1:50
  • Supplied in PBS (pH 7.2) containing <0.1% sodium azide and 2 mg/mL BSA
  • Species reactivity: Human, mouse, rat, and pig
  • Store at 4°C, protect from light, and do not freeze

Catalog No. 50-273-2812 Supplier Cell Signaling Technology Supplier No. 58036L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protein L is a recombinant protein that interacts with the κ light chain variable domain of immunoglobulins without disrupting paratope function. Conjugated to phycoerythrin (PE), this product is specifically validated for direct flow cytometric analysis of human cells expressing κ light chain-containing immunoglobulins, including Fab fragments and single chain variable fragments (scFvs). The protein has been genetically engineered to remove cell wall, cell membrane, and albumin-binding regions to enhance specificity.

  • Validated for direct flow cytometry (live)
  • Recommended dilution: 1:50
  • Supplied in PBS (pH 7.2) containing <0.1% sodium azide and 2 mg/mL BSA
  • Species reactivity: Human, mouse, rat, and pig
  • Store at 4°C, protect from light, and do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.