Learn More
Enzo Life Sciences Prestained Protein Ladder (3.5 - 245 kDa) (500µl)
SDP
Supplier:  Enzo Life Sciences ENZACC1310500
Encompass_Preferred
Enzo's Protein ladder is a three-color protein standard with 13 prestained proteins, covering a wide range molecular weights from 3.5 to 245 kDa. Proteins are covalently coupled with a blue chromophore except for two reference bands (one green and one red band at 25 kDa and 75 kDa respectively) when separated on SDS-PAGE (Tris-glycine buffer). Our Protein ladder is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes (PVDF, nylon, or nitrocellulose) and for approximating the size of proteins. The ladder is supplied in gel loading buffer and is ready to use. Do not heat, dilute or add reducing agent before loading. Formulation: Approximately 0.1~0.4 mg/ml of each protein in the buffer (20mM Tris-phosphate, pH 7.5 at 25°C), 2% SDS, 10mM Dithiothreitol, 3.6 M Urea, and 15% (v/v) Glycerol). Long Term Storage: -20°C. Applications: SDS-PAGE.
Catalog No. 50-201-3365