Learn More
Sino Biological Protein LASV LASV-G Biotin 20U
Catalog No. 5001074122
Encompass_Preferred
A DNA sequence encoding the Lassa virus strain Mouse/Sierra Leone/Josiah/1976 GP / Glycoprotein (P08669) (Thr59-Asp432) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-74122 Supplier Sino Biological Supplier No. 41180V49HB20
A DNA sequence encoding the Lassa virus strain Mouse/Sierra Leone/Josiah/1976 GP / Glycoprotein (P08669) (Thr59-Asp432) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed